missing translation for 'onlineSavingsMsg'
Learn More

PLX51107, MedChemExpress

Product Code. 16410767
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16410767

Brand: Medchem Express HY111422

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PLX51107 is a potent and selective BET inhibitor, with Kds of 1.6, 2.1, 1.7, and 5 nM for BD1 and 5.9, 6.2, 6.1, and 120 nM for BD2 of BRD2, BRD3, BRD4, and BRDT, respectively; PLX51107 also interacts with the bromodomains of CBP and EP300 (Kd, in the 100 nM range).

PLX51107 is a potent and selective BET inhibitor, with Kds of 1.6, 2.1, 1.7, and 5 nM for BD1 and 5.9, 6.2, 6.1, and 120 nM for BD2 of BRD2, BRD3, BRD4, and BRDT, respectively

Specifications

Chemical Name or Material PLX51107
CAS 1627929-55-8
Color White
Purity Grade Notes Research
Molecular Formula C26H22N4O3
Quantity 50 mg
Solubility Information DMSO : 75 mg/mL (171.05 mM; Need ultrasonic)
SMILES C[C@@H](C1=NC=CC=C1)N2C=C(C3=CC=C(C(O)=O)C=C3)C4=NC=C(C5=C(C)ON=C5C)C=C42
Molecular Weight (g/mol) 438.48
Formula Weight 438.48
Percent Purity 99.65%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.