missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies PLX4032
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
- PLX4032 is an ATP-competitive inhibitor of the serine/threonine kinase B-RAF proto-oncogene, with IC50 values of 31 and 100 nM for the wild-type and V600E mutant forms respectively (Khazak et al.; Sala et al.)
- Inhibits proliferation in colon, melanoma, and thyroid carcinoma cancer cell lines expressing B-RAF V600E, alone or in synergy with taxol, vinblastine, and oxaliplatin compounds (Khazak et al.)
- Suppresses MEK and ERK phosphorylation downstream of B-RAF in melanoma cells with mutations at the V600 position, correlated with antiproliferative effects (Joseph et al.; Yang et al.)
- Inhibits tumor growth in B-RAF V600E melanoma tumor xenograft models (Yang et al.)
Specifications
Specifications
| CAS | 918504-65-1 |
| Synonym | RG-7204; Ro 51-85426; Vemurafenib; Zelboraf™ |
| Quantity | 50 mg |
| Molecular Formula | C23H18ClF2N3O3S |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction