missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLVAP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115774
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PLVAP.
PLVAP involved in the formation of stomatal and fenestral diaphragms of caveolae. This protein may function in microvascular permeability.
Specifications
| PLVAP | |
| Polyclonal | |
| Unconjugated | |
| PLVAP | |
| FELS; Fenestrated endothelial-linked structure protein; fenestrated-endothelial linked structure protein; PV-1 protein; gp68; MECA32; MECA-32 antigen; plasmalemma vesicle associated protein; plasmalemma vesicle protein 1; Plasmalemma vesicle-associated protein; PLVAP; Pv1; PV-1; PV-1 protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 56765, 83483, 84094 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q91VC4, Q9BX97, Q9WV78 | |
| PLVAP | |
| A synthesized peptide derived from human PLVAP(Accession Q9BX97), corresponding to amino acid residues I390-S440. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction