missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLVAP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA526384
This item is not returnable.
View return policy
Description
PLVAP Polyclonal Antibody for Western Blot, Flow
PLVAP involved in the formation of stomatal and fenestral diaphragms of caveolae. This protein may function in microvascular permeability.
Specifications
| PLVAP | |
| Polyclonal | |
| Unconjugated | |
| PLVAP | |
| FELS; Fenestrated endothelial-linked structure protein; fenestrated-endothelial linked structure protein; PV-1 protein; gp68; MECA32; MECA-32 antigen; plasmalemma vesicle associated protein; plasmalemma vesicle protein 1; Plasmalemma vesicle-associated protein; PLVAP; Pv1; PV-1; PV-1 protein | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 83483 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 0.42 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9BX97 | |
| PLVAP | |
| KLH conjugated synthetic peptide between 214-242 amino acids from the central region of human PLVAP. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction