missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PluriSln 1

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Stearoyl-CoA desaturase 1 (SCD1) inhibitor. Used to selectively eliminate undifferentiated human pluripotent stem cells (hPSCs) from culture; induces ER stress, attenuates protein synthesis and induces apoptosis in hPSCs. Prevents teratoma formation in immunocompromised mice.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 213.24 |
| Purity | >99% |
| Molecular Weight (g/mol) | 213.24 |
| Chemical Name or Material | PluriSln 1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction