missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PluriSln 1
GREENER_CHOICE

Product Code. 30001619
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001619

Brand: Tocris Bioscience™ 4847/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SCD-1 inhibitor

Stearoyl-CoA desaturase 1 (SCD1) inhibitor. Used to selectively eliminate undifferentiated human pluripotent stem cells (hPSCs) from culture; induces ER stress, attenuates protein synthesis and induces apoptosis in hPSCs. Prevents teratoma formation in immunocompromised mice.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91396-88-2
Molecular Formula C12H11N3O

Specifications

Quantity 10 mg
Formula Weight 213.24
Purity >99%
Molecular Weight (g/mol) 213.24
Chemical Name or Material PluriSln 1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.