missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies PluriSIn-1
Description
- PluriSIn-1 is an N-acyl phenylhydrazine derivative that inhibits stearoyl-CoA desaturase, a key enzyme for lipid metabolism that is expressed in human pluripotent stem cells (Ben-David et al.)
- Selectively eliminates undifferentiated human embryonic stem (ES) and induced pluripotent stem (iPS) cells while sparing differentiated cells, and prevents teratoma formation in transplanted mice (Ben-David et al.)
- Induces apoptosis of Nanog-positive iPS cells in vitro, while leaving iPS cell-derived cardiomyocytes unaffected (Zhang et al.)
Specifications
Specifications
| CAS | 91396-88-2 |
| Synonym | N’-phenyl-hydrazine-Isonicotinic acid; NSC 14613 |
| Quantity | 50 mg |
| Molecular Formula | C12H11N3O |
| Purity | ≥95% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?