Learn More
Novus Biologicals™ Pluripotent Stem Cell Transcription Factor Antibody Pack
For Research applications
Brand:  Novus Biologicals™ NBP1-42823
Product Code. 18796133
  • £647.00 / Pack of 4
Includes:
  • 1.NB100-2379: OCT4 Antibody
  • 2.NB100-588: Nanog Antibody
  • 3.NB110-37235: SOX2 Antibody
Description
Description
Pluripotent Stem Cell Transcription Factor Polyclonal antibody specifically detects Pluripotent Stem Cell Transcription Factor in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
Affinity Purified
Mouse, Bovine, Chicken, Primate
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
NB100-58842 is specific for mouse Nanog protein. NB100-2379-OCT4 - Embryonic Stem Cell Marker NB110-37235 - SOX2 - Embryonic Stem Cell Marker
Store at 4C. Do not freeze.