missing translation for 'onlineSavingsMsg'
Learn More

Socorex™ Plunger for Acura™ Capillary Micropipettes

Product Code. 10618271
Change view
Click to view available options
Volume (Metric):
1 to 5 μL
100 to 200 μL
Color:
Red
Steel
Unit Size:
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Color Unit Size
10618271 1 to 5 μL Steel Pack of 5
10499820 100 to 200 μL Red Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10618271 Supplier Socorex™ Supplier No. 342.005
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ETFE-tipped stainless steel micropipette plunger. Socorex™ Plunger for Acura™ Capillary Micropipettes is designed with matching cap and tip color codes.

  • Stainless steel construction.
  • ETFE tip for chemical resistance.
  • Matching cap and tip color code.

Specifications

Volume (Metric) 1 to 5 μL
Color Steel
Quantity 5/Pk.
Product Type Spare Plunger with Glass Capillaries
For Use With (Equipment) Acura Capillar 846 MicroPipette
Material Stainless Steel, ETFE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.