missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLUNC Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-26120
This item is not returnable.
View return policy
Description
PLUNC Polyclonal specifically detects PLUNC in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| PLUNC | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| BPIFA1 | |
| Peptide with sequence C-NEVLRGLDITLVHD corresponding to C-Terminus according to NP_057667.1. | |
| 0.1 mg | |
| Immunology | |
| 51297 | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:64000 | |
| bA49G10.5, ligand-binding protein RYA3, LPLUNC3, Lung-specific protein X, LUNXNASG, Nasopharyngeal carcinoma-related protein, Palate lung and nasal epithelium clone protein, palate, lung and nasal epithelium associated, palate, lung and nasal epithelium carcinoma associated, protein Plunc, Secretory protein in upper respiratory tracts, SPLUNC1SPURT, tracheal epithelium enriched protein, Tracheal epithelium-enriched protein, Von Ebner protein Hl | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Reported variants represent identical protein: NP_057667.1, NP_001230122.1, NP_570913.1 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction