missing translation for 'onlineSavingsMsg'
Learn More

Plumbagin, MedChemExpress

Product Code. 16484917
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16484917 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16484917 Supplier Medchem Express Supplier No. HYN1497
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Plumbagin (2-Methyljuglone) is a naphthoquinone isolated from Plumbago zeylanica L, exhibits anticancer and antiproliferative activities.

Plumbagin (2-Methyljuglone) is a naphthoquinone isolated from Plumbago zeylanica L, exhibits anticancer and antiproliferative activities.

Specifications

Chemical Name or Material Plumbagin
CAS 481-42-5
Color Orange
Purity Grade Notes Research
Molecular Formula C11H8O3
Quantity 50 mg
Synonym 2-Methyljuglone
Solubility Information DMSO : ≥ 150 mg/mL (797.11 mM)
SMILES O=C1C(C)=CC(C2=C1C=CC=C2O)=O
Molecular Weight (g/mol) 188.18
Formula Weight 188.18
Percent Purity 98.16%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.