missing translation for 'onlineSavingsMsg'
Learn More

Labcon Plug Style Caps for 12 x 75 mm Culture Tubes

Product Code. 17265704
Click to view available options
Color:
Assorted
Blue
Green
Natural
Orange
Purple
Red
White
Yellow
Unit Size:
Pack of 1000
Pack of 500
This item is not returnable. View return policy

Product Code. 17265704

Brand: Labcon 33028700009

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Cap includes flutes to securely grip sides of tubes

  • Fluted Plug Style Cap
  • Plug cap for culture tubes
  • Fits 12 x 75 mm culture tubes
  • Cap includes flutes to securely grip sides of tubes
  • Endotoxin Free (non-pyrogenic)
  • Polyethylene USP Class VI resin
  • Made in USA

Specifications

Certifications/Compliance ISO 9001:2015; FDA CFR21 GMP
Closure Size 13 mm
Pre-sterilized No
Product Type Tube Plugs
Autoclavable Not Autoclavable
Material Polyethylene
Color Assorted
For Use With (Application) Centrifugation; For Cell Culture and General Laboratory Use
Clean Claims ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
Length (Metric) 75 mm
Sterility Non-sterile
For Use With (Equipment) Tubes
Disposable Single-use
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis