missing translation for 'onlineSavingsMsg'
Learn More

PLSCR4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-85501-0.1ML

Product Code. 18253819

  • £361.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PLSCR4 Polyclonal specifically detects PLSCR4 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PLSCR4
Polyclonal
PBS, 2% Sucrose
Rabbit
Immunogen affinity purified
RUO
57088
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
Ca(2+)-dependent phospholipid scramblase 4, cell growth inhibiting protein 43, Cell growth-inhibiting gene 43 protein, phospholipid scramblase 4, PL scramblase 4, TRA1
The immunogen is a synthetic peptide directed towards the C-terminal region of Human PLSCR4
0.1 mL
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.