missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLSCR4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA535951
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (87%), Rat (88%), Rhesus Monkey (98%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May mediate accelerated ATP-independent bidirectional transbilayer migration of phospholipids upon binding calcium ions that results in a loss of phospholipid asymmetry in the plasma membrane. May play a central role in the initiation of fibrin clot formation, in the activation of mast cells and in the recognition of apoptotic and injured cells by the reticuloendothelial system.
Specifications
| PLSCR4 | |
| Polyclonal | |
| Unconjugated | |
| PLSCR4 | |
| AV245873; Ca(2+)-dependent phospholipid scramblase 4; cell growth inhibiting protein 43; Cell growth-inhibiting gene 43 protein; GIG43; Phospholipid scramblase 4; PL scramblase 4; Pls4; PLSCR4; TRA1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 235527, 57088 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P58196, Q9NRQ2 | |
| PLSCR4 | |
| Recombinant protein encompassing a sequence within the center region of human PLSCR4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction