missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLSCR3 Recombinant Rabbit Monoclonal Antibody (24GB1345)

Rabbit Recombinant Monoclonal Antibody
£172.00 - £488.00
Specifications
| Antigen | PLSCR3 |
|---|---|
| Clone | 24GB1345 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275235
![]() |
Invitrogen™
MA557582 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30274921
|
Invitrogen™
MA557583 |
20 μL |
£172.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PLSCR3 Recombinant Monoclonal Antibody for Western Blot, Flow
May mediate accelerated ATP-independent bidirectional transbilayer migration of phospholipids upon binding calcium ions that results in a loss of phospholipid asymmetry in the plasma membrane. May play a central role in the initiation of fibrin clot formation, in the activation of mast cells and in the recognition of apoptotic and injured cells by the reticuloendothelial system. Seems to play a role in apoptosis, through translocation of cardiolipin from the inner to the outer mitochondrial membrane which promotes BID recruitment and enhances tBid-induced mitochondrial damages. [UniProt]Specifications
| PLSCR3 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| Plscr3 | |
| 2210403O21Rik; 2610037N06Rik; Ca 2+ dependent phospholipid scramblase 3; ca(2+)-dependent phospholipid scramblase 3; ESTM3; LOW QUALITY PROTEIN: phospholipid scramblase 3; LOW QUALITY PROTEIN: phospholipid scramblase 3; phospholipid scramblase 3; phospholipid scramblase 3; PL scramblase 3; Pls3; Plscr3; Unknown (protein for MGC:138060); X83310 | |
| Plscr3 | |
| Primary | |
| Affinity chromatography |
| 24GB1345 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6QBQ4, Q9JIZ9, Q9NRY6 | |
| 360549, 57048, 70310 | |
| A synthesized peptide derived from human PLSCR3 (aa 1-45). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title