missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLSCR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | PLSCR1 |
|---|---|
| Concentration | 0.46 mg/mL |
| Applications | Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16818763
|
Proteintech
11582-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16808763
|
Proteintech
11582-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The calcium-dependent plasma membrane protein phospholipid scramblase 1 (PLSCR1) contributes to the transbilayer movement of phosphatidylserine and other membrane phospholipids upon influx of calcium into the cytosol. This movement results in plasma membrane phospholipid remodelling and surface exposure of phosphatidylserine in injured or apoptotic cells, which leads to cell death. Interferons and other cytokines induce expression of PLSCR1, implying that PLSCR1 also functions in cytokine signaling pathways. EGF stilmulation results in tyrosine phosphorylation of PLSCR1 on tyrosines 69 and 74, which allows it to interact with Shc, and thereby connecting Src kinase activation to stimulation of the EGF receptor.Specifications
| PLSCR1 | |
| Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human, Rat | |
| O15162, P58195, Q9JJ00 | |
| 117540, 22038, 5359 | |
| PLSCR1 Fusion Protein Ag2172 | |
| Primary | |
| -20°C | |
| PLSCR1 |
| 0.46 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| MMTRA1B, phospholipid scramblase 1, PL scramblase 1, PLSCR1 | |
| PLSCR1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title