Learn More
Invitrogen™ PLRG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528418
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Raji. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (100%), Pig (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Plrg1 is a protein coding gene involved in pre-mRNA splicing as component of the spliceosome. Component of the PRP19-CDC5L complex that forms an integral part of the spliceosome and is required for activating pre-mRNA splicing.
Specifications
| PLRG1 | |
| Polyclonal | |
| Unconjugated | |
| PLRG1 | |
| AA958940; C80566; Cwc1; MGC110980; MGC137269 protein; pleiotropic regulator 1; pleiotropic regulator 1 (PRL1 homolog, Arabidopsis); pleiotropic regulator 1 homolog; pleiotropic regulator 1, PRL1 homolog (Arabidopsis); pleiotropic regulator 1-like protein; PLRG1; PRL1; PRP46; PRPF46; Tango4; transport and golgi organization 4 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 53317, 5356 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 10% glycerol and 0.01% thimerosal; pH 7 | |
| O43660, Q922V4 | |
| PLRG1 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human PLRG1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.