missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLRG1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557468
This item is not returnable.
View return policy
Description
Immunogen sequence: NTLTVNSDGV LVSGADNGTM HLWDWRTGYN FQRVHAAVQP GSLDSESGIF ACAFDQSESR LLTAEADKTI KVYREDDTAT EETHPVS Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
Plrg1 is a protein coding gene involved in pre-mRNA splicing as component of the spliceosome. Component of the PRP19-CDC5L complex that forms an integral part of the spliceosome and is required for activating pre-mRNA splicing.
Specifications
| PLRG1 | |
| Polyclonal | |
| Unconjugated | |
| PLRG1 | |
| AA958940; C80566; Cwc1; MGC110980; MGC137269 protein; pleiotropic regulator 1; pleiotropic regulator 1 (PRL1 homolog, Arabidopsis); pleiotropic regulator 1 homolog; pleiotropic regulator 1, PRL1 homolog (Arabidopsis); pleiotropic regulator 1-like protein; PLRG1; PRL1; PRP46; PRPF46; Tango4; transport and golgi organization 4 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 53317, 5356, 60376 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43660, Q922V4, Q9WUC8 | |
| PLRG1 | |
| Recombinant protein corresponding to Human PLRG1. Recombinant protein control fragment (Product #RP-96577). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction