missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PLpro inhibitor 6

Product Code. p-200039980
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18064992

Brand: Tocris Bioscience™ 7357/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

SARS-CoV and SARS-CoV-2 PLpro inhibitor

PLpro inhibitor 6 is a SARS-CoV and SAR-CoV-2 PLpro inhibitor (IC50 values are 2.6 and 5.0 μM, respectively). Inhibits SARS-CoV and SARS-CoV-2 infection of Vero E6 cells in vitro (EC50 values are 13.1 and 21 μM, respectively).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-(Acetylamino)-2-methyl-N-[(1 R)-1-(1-naphthalenyl)ethyl]benzamide
CAS 1093070-14-4
Quantity 5 mg
Target Papain-like Protease Inhibitors
Molecular Formula C22H22N2O2
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.