missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PLpro inhibitor 6
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
PLpro inhibitor 6 is a SARS-CoV and SAR-CoV-2 PLpro inhibitor (IC50 values are 2.6 and 5.0 μM, respectively). Inhibits SARS-CoV and SARS-CoV-2 infection of Vero E6 cells in vitro (EC50 values are 13.1 and 21 μM, respectively).
Specifications
Specifications
| Chemical Name or Material | 5-(Acetylamino)-2-methyl-N-[(1 R)-1-(1-naphthalenyl)ethyl]benzamide |
| CAS | 1093070-14-4 |
| Quantity | 5 mg |
| Target | Papain-like Protease Inhibitors |
| Molecular Formula | C22H22N2O2 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction