missing translation for 'onlineSavingsMsg'
Learn More

PLPP4/PPAPDC1A Antibody - BSA Free, Novus Biologicals™

Product Code. 18337580 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18337580 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18337580 Supplier Novus Biologicals Supplier No. NBP281931

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PLPP4/PPAPDC1A Polyclonal specifically detects PLPP4/PPAPDC1A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLPP4/PPAPDC1A
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 20 μg/mL
Formulation PBS
Gene Alias Diacylglycerol Pyrophosphate Like 2, DPPL2, Phosphatidate Phosphatase PPAPDC1A, Phosphatidic Acid Phosphatase Type 2 Domain Containing 1A, Phospholipid Phosphatase 4, PPAPDC1
Host Species Rabbit
Immunogen PPAPDC1A antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human PPAPDC1A. The immunogen is located within the last 50 amino acids of PPAPDC1A. Amino Acid Squence: ASLKKEERPTADSA
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196051
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.