missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLP1 Monoclonal Antibody (plpc1)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA180034
This item is not returnable.
View return policy
Description
Predicted to react with all mammals based on sequence homology. Mouse anti myelin proteolipid protein antibody, clone plpc1 recognizes myelin proteolipid protein (PLP) in many mammalian species (Stoffel et al. 1985).
PLP1 is a transmembrane proteolipid protein that is the predominant myelin protein present in the central nervous system. It may play a role in the compaction, stabilization, and maintenance of myelin sheaths, as well as in oligodendrocyte development and axonal survival. Mutations in this gene cause X-linked Pelizaeus-Merzbacher disease and spastic paraplegia type 2. Alternatively spliced transcript variants encoding distinct isoforms or having different 5' UTRs, have been identified for this gene.This gene encodes a transmembrane proteolipid protein that is the predominant myelin protein present in the central nervous system. It may play a role in the compaction, stabilization, and maintenance of myelin sheaths, as well as in oligodendrocyte development and axonal survival. Mutations in this gene cause X-linked Pelizaeus-Merzbacher disease and spastic paraplegia type 2. Alternatively spliced transcript variants encoding distinct isoforms or having different 5' UTRs, have been identified for this gene.
Specifications
| PLP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with <0.1% sodium azide | |
| P04116, P23294, P60201, P60202, P60203 | |
| PLP1 | |
| Synthetic peptide GRGTKF corresponding to C terminal region of myelin proteolipid protein. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey, Bovine | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| plpc1 | |
| Unconjugated | |
| PLP1 | |
| DM20; GPM6C; HLD1; jimpy; jp; lipophilin; lipophilin (aa 129 to 276); major myelin proteolipid protein; MMPL; msd; Myelin proteolipid protein; myelin synthesis deficiency; PLP; PLP/DM20; Plp1; PMD; proteolipid; proteolipid protein; proteolipid protein (myelin) 1; Proteolipid protein (Pelizaeus-Merzbacher disease spastic paraplegia 2 uncomplicated); Proteolipid protein (Pelizaeus-Merzbacher disease, spastic paraplegia 2, uncomplicated); proteolipid protein 1; proteolipid protein 1 (Pelizaeus-Merzbacher disease, spastic paraplegia 2, uncomplicated); proteolipid protein, lipophilin; rsh; rump shaker; SPG2 | |
| Mouse | |
| Protein G | |
| RUO | |
| 18823, 24943, 281410, 481002, 5354 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction