missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Plozalizumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558469
This item is not returnable.
View return policy
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Plozalizumab has been investigated for the treatment of Atherosclerosis. Plozalizumab, a novel humanized monoclonal antibody, specifically targets CCR2 chemokine receptors found on the surface of certain white blood cells including macrophages and monocytes. Preclinical studies suggest that CCR2 plays an important role in the trafficking of monocytes and macrophages to sites of inflammation. The recruitment of macrophages to the arterial wall is believed to be a critical step in the development of atherosclerosis.
Specifications
| Plozalizumab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| hu1D9; MLN-1202 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction