missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLOD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548435
This item is not returnable.
View return policy
Description
PLOD3 Polyclonal Antibody for Western Blot
Forms hydroxylysine residues in -Xaa-Lys-Gly- sequences in collagens. These hydroxylysines serve as sites of attachment for carbohydrate units and are essential for the stability of the intermolecular collagen cross-links.
Specifications
| PLOD3 | |
| Polyclonal | |
| Unconjugated | |
| PLOD3 | |
| AI414586; bone protein I (BP-I); Galactosylhydroxylysine-glucosyltransferase; LH3; lysine hydroxylase 3; lysyl hydroxlase 3; lysyl hydroxylase 2; lysyl hydroxylase 3; Multifunctional procollagen lysine hydroxylase and glycosyltransferase LH3; Plod3; Procollagen galactosyltransferase; Procollagen glucosyltransferase; Procollagen glycosyltransferase; procollagen-lysine, 2-oxoglutarate 5-dioxygenase 3; procollagen-lysine,2-oxoglutarate 5-dioxygenase 3 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 8985 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O60568 | |
| PLOD3 | |
| KLH conjugated synthetic peptide between 78-105 amino acids from the N-terminal region of human PLOD3. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction