missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLOD2 Mouse anti-Human, Clone: 1H9E1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66342-1-IG-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a membrane-bound homodimeric enzyme that is localized to the cisternae of the rough endoplasmic reticulum. The enzyme (cofactors iron and ascorbate) catalyzes the hydroxylation of lysyl residues in collagen-like peptides. The resultant hydroxylysyl groups are attachment sites for carbohydrates in collagen and thus are critical for the stability of intermolecular crosslinks. Some patients with Ehlers-Danlos syndrome type VIB have deficiencies in lysyl hydroxylase activity. Mutations in the coding region of this gene are associated with Bruck syndrome. Alternative splicing results in multiple transcript variants encoding different isoforms.Specifications
| PLOD2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O00469 | |
| PLOD2 | |
| PLOD2 Fusion Protein Ag5779 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgA |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1H9E1 | |
| Unconjugated | |
| PLOD2 | |
| LH2, Lysyl hydroxylase 2, PLOD2, TLH | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5352 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction