missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLOD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561892
This item is not returnable.
View return policy
Description
Immunogen sequence: YISNIYLIKG SALRGELQSS DLFHHSKLDP DMAFCANIRQ QDVFMFLTNR HTLGHLLSLD SYRTTHLHND LWEVFSN Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
Forms hydroxylysine residues in -Xaa-Lys-Gly- sequences in collagens. These hydroxylysines serve as sites of attachment for carbohydrate units and are essential for the stability of the intermolecular collagen cross-links.
Specifications
| PLOD1 | |
| Polyclonal | |
| Unconjugated | |
| PLOD1 | |
| 2410042F05Rik; AI854890; AV007929; EDS6; LH; LH1; LLH; lysine hydroxylase; lysyl hydroxlase 1; Lysyl hydroxylase 1; Plod; Plod1; procollagen-lysine 1, 2-oxoglutarate 5-dioxygenase 1; procollagen-lysine, 2-oxoglutarate 5-dioxygenase 1; procollagen-lysine,2-oxoglutarate 5-dioxygenase 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5351 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q02809 | |
| PLOD1 | |
| Recombinant protein corresponding to Human PLOD1. Recombinant protein control fragment (Product #RP-101122). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction