missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLK4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12952-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the polo family of serine/threonine protein kinases. The protein localizes to centrioles, complex microtubule-based structures found in centrosomes, and regulates centriole duplication during the cell cycle.Specifications
| PLK4 | |
| Polyclonal | |
| Unconjugated | |
| PLK4 | |
| PLK 4, PLK4, Polo like kinase 4, SAK, STK18 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10733, 20873, 310344 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| B2GUY1, O00444, Q64702 | |
| Plk4 | |
| PLK4 Fusion Protein Ag3605 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction