Learn More
Invitrogen™ Human PLK3, GST Tag Recombinant Protein
Description
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Catalytic
Specifications
Specifications
| Accession Number | Q9H4B4 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 |
| Gene ID (Entrez) | 1263 |
| Molecular Weight (g/mol) | 59.7 kDa |
| Name | Human PLK3, GST Tag |
| Quantity | 1 mg |
| Regulatory Status | RUO - research use only |
| Gene Alias | CNK; cytokine inducible kinase; cytokine-inducible serine/threonine-protein kinase; FGF-inducible kinase; Fnk; PLK3; PLK-3; polo like kinase 3; polo-like kinase 3; PRK; Proliferation-related kinase; serine/threonine-protein kinase PLK3; serine-threonine kinase |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.