missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLK3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113431
This item is not returnable.
View return policy
Description
PLK3 Polyclonal Antibody for Western Blot, IHC (P), ELISA
PLK3 is a member of the polo family of serine/threonine kinases. PLK3 is involved in the regulation of M phase in the cell cycle. CNK contains both a catalytic domain and a putative regulatory domain. It may play a role in regulation of cell cycle progression and tumorigenesis.
Specifications
| PLK3 | |
| Polyclonal | |
| Unconjugated | |
| PLK3 | |
| CNK; cytokine inducible kinase; cytokine-inducible serine/threonine-protein kinase; FGF-inducible kinase; Fnk; PLK3; PLK-3; polo like kinase 3; polo-like kinase 3; PRK; Proliferation-related kinase; serine/threonine-protein kinase PLK3; serine-threonine kinase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1263, 58936 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9H4B4, Q9R011 | |
| PLK3 | |
| Recombinant Human Serine/threonine-protein kinase PLK3 protein (487-646AA). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction