missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PLK1, His Tag Recombinant Protein

Product Code. 15458443
Click to view available options
Quantity:
10 μg
100 μg
Unit Size:
100µg
10µg
This item is not returnable. View return policy

Product Code. 15458443

Brand: Invitrogen™ PR5304B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.

PLK1 is a serine/threonine kinase that is thought to function in cell mitosis and regarded as a marker of cell proliferation. It has been found to play a critical role in carcinogenesis. Polo-like kinase proteins acts by binding and phosphorylating proteins are that already phosphorylated on a specific motif recognized by the POLO box domains. Phosphorylates BORA, BUB1B/BUBR1, CCNB1, CDC25C, CEP55, ECT2, ERCC6L, FBXO5/EMI1, FOXM1, KIF20A/MKLP2, CENPU, NEDD1, NINL, NPM1, NUDC, PKMYT1/MYT1, KIZ, PPP1R12A/MYPT1, PRC1, RACGAP1/CYK4, SGOL1, STAG2/SA2, TEX14, TOPORS, p73/TP73, TPT1 and WEE1. Plays a key role in centrosome functions and the assembly of bipolar spindles by phosphorylating KIZ, NEDD1 and NINL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P53350
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 20mM HEPES with 0.05% Triton X-100, 50% glycerol, 5mM DTT, 0.5mM EDTA, 150mM NaCl, 0.5mM EGTA and no preservative; pH 7.5
Gene ID (Entrez) 5347
Molecular Weight (g/mol) 73.3 kDa
Name Human PLK1, His Tag
pH Range 7.5
Purification Method Purified
Quantity 100 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias cell cycle regulated protein kinase; Plk; PLK 1; Plk1; PLK-1; plk1.S; plx1; polo (Drosophia)-like kinase; polo like kinase; polo like kinase 1; polo like kinase 1 S homeolog; polo-like kinase; polo-like kinase 1; polo-like kinase homolog; polo-like protein kinase; serine/threonine-protein kinase 13; Serine/threonine-protein kinase PLK1; STPK13; STPK14; Unknown (protein for MGC:133927); XELAEV_18047692mg
Common Name PLK1
Gene Symbol Plk1
KinaseFamily PLK Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Kinase Group Ser⁄Thr Kinases: Other
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Cell Cycle & Proliferation
Purity or Quality Grade 85% by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.