Learn More
Eurofins DiscoverX Plk1 Protein, Active
Description
Polo-like kinases (PLKs) are a family of serine/threonine kinases related to the polo gene product of Drosophila melanogaster, originally identified from a mitotic mutant that displayed abnormal spindle poles. PLKs play essential roles in several stages of mitosis and cytokinesis with a dynamic pattern of localization to centrosomes, kinetochores and central spindle structures during the cell cycle. PLK1 has been shown to be overexpressed in many human tumours, such as non-small cell lung cancer, squamous cell carcinomas of the head and neck, melanomas, oropharyngeal carcinomas, and ovarian and endometrial carcinomas.
GenBank NM_005030
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 69.3 kDa |
| Quantity | 250 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant protein expressed in Sf21 insect cells |
| Protein Form | Full-length |
| Purity or Quality Grade | ≥72% |
| Protein | Plk1, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.