missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLK1 (PHO T210 Mouse anti-Human, Clone: 2A3, Abnova™
Mouse monoclonal antibody raised against synthetic phosphopeptide of human PLK1.
Brand: Abnova MAB12911.50ug
This item is not returnable.
View return policy
Specifications
| PLK1 (PHO T210 | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (15 ug/mL) Western Blot The optimal working dilution should be determined by the end user. | |
| PLK/STPK13 | |
| Mouse | |
| Affinity purification | |
| RUO | |
| 5347 | |
| Store at 4°C. Do not freeze. | |
| IgG1 |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 2A3 | |
| Mouse monoclonal antibody raised against synthetic phosphopeptide of human PLK1. | |
| In PBS, pH 7.2 (0.09% sodium azide). | |
| PLK1 | |
| Synthetic phosphopeptide corresponding to residues surrounding T210 of human PLK1. | |
| 50 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction