missing translation for 'onlineSavingsMsg'
Learn More

PLK1 Antibody (13E8), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NB100-74502-0.025mg

 View more versions of this product

Product Code. 18537163

  • £169.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PLK1 Monoclonal specifically detects PLK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PLK1
Monoclonal
1 mg/mL
Western Blot 1:1000, Immunohistochemistry 1:20, Immunocytochemistry/Immunofluorescence 1:20, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:20
P53350
PLK1
Recombinant protein containing residues 300-400 of human PLK1.
0.025 mg
Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Repair, Mitotic Regulators, Stem Cell Markers
5347
Human
Purified
Western Blot
1300000000
Unconjugated
PBS with 1mg/ml BSA with 0.05% Sodium Azide
cell cycle regulated protein kinase, EC 2.7.11, EC 2.7.11.21, PLK, PLK-1, polo (Drosophia)-like kinase, polo like kinase, polo-like kinase 1polo-like kinase (Drosophila), Serine/threonine-protein kinase 13, serine/threonine-protein kinase PLK1, STPK13
Mouse
Protein G purified
RUO
Primary
Detects PLK1 from human, mouse, and rat samples.
Store at -20°C. Avoid freeze/thaw cycles.
IgG2b
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only