missing translation for 'onlineSavingsMsg'
Learn More

Plinabulin, MedChemExpress

Product Code. 16496068
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496068 10 mg 10mg
16486466 100 mg 100mg
16482207 50 mg 50mg
16448677 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16496068 Supplier Medchem Express Supplier No. HY14444
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Plinabulin (NPI-2358) is a vascular disrupting agen (VDA) against tubulin-depolymerizing with an IC50 of 9.8 nM against HT-29 cells. Plinabulin binds the colchicine binding site of β-tubulin preventing polymerization and has potent inhibitory to tumor cells.

Plinabulin (NPI-2358) is a vascular disrupting agen (VDA) against tubulin-depolymerizing with an IC50 of 9.8 nM against HT-29 cells. Plinabulin binds the colchicine binding site of β-tubulin preventing polymerization and has potent inhibitory to tumor cells.

Specifications

Chemical Name or Material Plinabulin
CAS 714272-27-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H20N4O2
Quantity 10 mg
Synonym NPI-2358
Solubility Information DMSO : 50 mg/mL (148.64 mM; Need ultrasonic)
SMILES O=C(N/C1=C\C2=C(NC=N2)C(C)(C)C)/C(NC1=O)=C/C3=CC=CC=C3
Molecular Weight (g/mol) 336.39
Formula Weight 336.39
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.