missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLIN4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113147
This item is not returnable.
View return policy
Description
PLIN4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Specifications
| PLIN4 | |
| Polyclonal | |
| Unconjugated | |
| PLIN4 | |
| adipocyte protein S3-12; KIAA1881; perilipin 4; perilipin-4; PLIN4; S3-12 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 729359 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q96Q06 | |
| PLIN4 | |
| Recombinant Human Perilipin-4 protein (308-418AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction