missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLGRKT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5107217
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 92%; Dog: 85%; Guinea Pig: 85%; Horse: 85%; Human: 100%; Mouse: 85%; Rabbit: 85%; Rat: 92%.
The exact function of C9orf46 remains unknown.
Specifications
| PLGRKT | |
| Polyclonal | |
| Unconjugated | |
| PLGRKT | |
| 1110007H22Rik; 5033414D02Rik; AD025; AI852040; C9orf46; MDS030; Plasminogen receptor (KT); plasminogen receptor with a C-terminal lysine; plasminogen receptor, C-terminal lysine transmembrane protein; Plg-R(KT); PLGRKT; Plg-RKT; RGD1306839; transmembrane protein C9orf46; transmembrane protein C9orf46 homolog | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55848 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q9HBL7 | |
| PLGRKT | |
| Synthetic peptide directed towards the middle region of human C9orf46 (aa 100-147). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction