missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLGF Antibody (S08-9C2), Novus Biologicals™
Rabbit Monoclonal Antibody
£328.00 - £501.00
Specifications
| Antigen | PLGF |
|---|---|
| Clone | S08-9C2 |
| Concentration | 0.3 mg/mL |
| Dilution | Western Blot 1:2000 |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18620935
|
Novus Biologicals
NBP3-20019-50ul |
50 μg |
£328.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18672415
|
Novus Biologicals
NBP3-20019-100ul |
100 μg |
£501.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PLGF Monoclonal antibody specifically detects PLGF in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| PLGF | |
| 0.3 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 5228 | |
| IgG | |
| Affinity purified |
| S08-9C2 | |
| Western Blot 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| D12S1900, PGFL, placenta growth factor, placental growth factor, placental growth factor, vascular endothelial growth factor-related protein, PlGF, PlGF-2, PLGFplacental growth factor-like, SHGC-10760 | |
| A synthetic peptide of human PLGF (Uniprot # P49763) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title