missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLGF Antibody (37203.111), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-10966AF532
This item is not returnable.
View return policy
Description
PLGF Monoclonal specifically detects PLGF in Human samples. It is validated for Western Blot, ELISA.
Specifications
| PLGF | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PGF | |
| Purified recombinant human placenta growth factor expressed in E. coli. | |
| Primary | |
| In immunoblotting, the shows no cross-reactivity with recombinanthuman VEGF, PDGFAA, PDGFAB, and PDGFBB. | |
| Store at 4°C in the dark. |
| Western Blot, ELISA | |
| 37203.111 | |
| Western Blot, ELISA | |
| D12S1900, PGFL, placenta growth factor, placental growth factor, placental growth factor, vascular endothelial growth factor-related protein, PlGF, PlGF-2, PLGFplacental growth factor-like, SHGC-10760 | |
| Mouse | |
| 0.1 mL | |
| 5228 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction