missing translation for 'onlineSavingsMsg'
Learn More

PLGA (50:50), MedChemExpress

Product Code. 16452399
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16452399 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16452399 Supplier Medchem Express Supplier No. HYB2247
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress PLGA (poly(lactic-co-glycolic acid)) (50:50) is a copolymer of poly lactic acid (PLA) and poly glycolic acid (PGA) which can be used to fabricate devices for drug delivery and tissue engineering applications.

PLGA (poly(lactic-co-glycolic acid)) (50:50) is a copolymer of poly lactic acid (PLA) and poly glycolic acid (PGA) which can be used to fabricate devices for drug delivery and tissue engineering applications.

Specifications

Chemical Name or Material PLGA (50:50)
Color White
CAS 34346-01-5
Physical Form Solid
Purity Grade Notes Research
Synonym poly(lactic-co-glycolic acid) (50:50)
Solubility Information DMSO : 25 mg/mL (Need ultrasonic) ∣H2O : 0.1 mg/mL (Need ultrasonic)
Quantity 1 g
SMILES O=C(O)C(C)OC(CO[H])=O.[x].[y]
Grade Research

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.