missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plexin D1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB300-961
This item is not returnable.
View return policy
Description
Plexin D1 Polyclonal specifically detects Plexin D1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA.
Specifications
| Plexin D1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| KIAA0620MGC75353, PLEXD1, plexin D1, plexin-D1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 23129 | |
| Human, Mouse | |
| IgG |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA | |
| 0.5 mg/mL | |
| Flow Cytometry 10 ug/mL, Immunocytochemistry/Immunofluorescence 10 ug/mL, Peptide ELISA Detection limit 1:16000 | |
| NP_055918. | |
| PLXND1 | |
| Peptide with sequence C-LAEPKKSHRQSH corresponding to internal region according to NP_055918.2. | |
| 0.1 mg | |
| Primary | |
| Expected from sequence similarity: Dog, Pig, Cow. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction