missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plexin D1 Antibody (752829), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4160V
This item is not returnable.
View return policy
Description
Plexin D1 Monoclonal antibody specifically detects Plexin D1 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Plexin D1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 23129 | |
| Store at 4C in the dark. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 752829 | |
| Flow Cytometry, CyTOF-ready | |
| KIAA0620MGC75353, PLEXD1, plexin D1, plexin-D1 | |
| Mouse myeloma cell line NS0-derived recombinant human Plexin D1, Leu47-Ala1271, Accession # Q9Y4D7 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction