missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Plexin C1 Antibody (3C10B2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27144-0.025ml
This item is not returnable.
View return policy
Description
Plexin C1 Monoclonal antibody specifically detects Plexin C1 in Human samples. It is validated for Flow Cytometry,ELISA,Western Blot
Specifications
| Plexin C1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 | |
| CD232, CD232 antigen, plexin (semaphorin receptor), plexin C1, PLXN-C1, receptor for viral semaphorin protein, receptor for virally-encoded semaphorin, VESPRplexin-C1, Virus-encoded semaphorin protein receptor | |
| Purified recombinant fragment of human Plexin C1 (AA: extra 35-234) expressed in E. Coli. | |
| 0.025 mL | |
| Growth and Development, Neuronal Cell Markers | |
| 10154 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Flow Cytometry, ELISA, Western Blot | |
| 3C10B2 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction