missing translation for 'onlineSavingsMsg'
Learn More

Plexin C1 Antibody (1A12), Novus Biologicals™

Product Code. 18359709 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18359709 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18359709 Supplier Novus Biologicals Supplier No. H00010154M06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Plexin C1 Monoclonal antibody specifically detects Plexin C1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Plexin C1
Applications ELISA
Classification Monoclonal
Clone 1A12
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_005752
Gene Alias CD232, CD232 antigen, plexin (semaphorin receptor), plexin C1, PLXN-C1, receptor for viral semaphorin protein, receptor for virally-encoded semaphorin, VESPRplexin-C1, Virus-encoded semaphorin protein receptor
Host Species Mouse
Immunogen PLXNC1 (NP_005752, 250 a.a. ~ 348 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PYNYTSGAATGWPSMARIAQSTEVLFQGQASLDCGHGHPDGRRLLLSSSLVEALDVWAGVFSAAAGEGQERRSPTTTALCLFRMSEIQARAKRVSWDFK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10154
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.