missing translation for 'onlineSavingsMsg'
Learn More

Plexin B3 Antibody [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30737142 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737142 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737142 Supplier Novus Biologicals Supplier No. AF6879JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Plexin B3 Polyclonal antibody specifically detects Plexin B3 in Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Plexin B3
Applications Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF
Classification Polyclonal
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Gene Alias PLEXB3, plexin B3, plexin-B3, PLEXR
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Plexin B3, His25-Gln1235, Accession # Q9QY40
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5365
Target Species Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.