missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plexin B2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10602-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody raised against 1542-1841aa of human plexin B2 can recognize precursor and beta subunit of plexin B2.
Plexin-B2 is a member of the Semaphorin receptor family. The semaphorin family consists of membrane and secreted proteins involved in attraction and repulsion of neuronal guidance cues. Plexin-B1 is a transmembrane protein that can be cleaved coverting the single chain presursor into a heterodimeric receptor. It has been shown to bind Sema4D/CD100 and is involved in axon development in mouse embryos. Sema4D also plays a role in the immune system interacting with CD72. Plexin-B2′s possible role in the immune system has not been characterized.Especificaciones
| Plexin B2 | |
| Polyclonal | |
| Unconjugated | |
| PLXNB2 | |
| KIAA0315, MM1, Nbla00445, PLEXB2, Plexin B2, PLXNB2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 140570, 23654, 315217 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.55 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| B2RXS4, O15031 | |
| Plxnb2 | |
| Plexin B2 Fusion Protein Ag0941 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido