missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plexin B1 Antibody (559830), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB37491C
This item is not returnable.
View return policy
Description
Plexin B1 Monoclonal antibody specifically detects Plexin B1 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Plexin B1 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5364 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| 559830 | |
| Flow Cytometry, CyTOF-ready | |
| plexin B1, SEP | |
| NS0 mouse myeloma cell line transfected with human Plexin B1, Accession # O43157 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction