missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plexin A3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93993-0.02ml
This item is not returnable.
View return policy
Description
Plexin A3 Polyclonal antibody specifically detects Plexin A3 in Mouse samples. It is validated for Western Blot
Specifications
| Plexin A3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| HSSEXGENE, plexin A3, Plexin-4, plexin-A3, PLXN3, PLXN4, Semaphorin receptor SEX, Sex chromosome X transmembrane protein of HGF receptor family 3, SEXPLEXIN-A3,6.3, XAP-6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 950-1150 of human PLXNA3 (NP_059984.3). GTRLTISGSSLDAGSRVTVTVRDSECQFVRRDAKAIVCISPLSTLGPSQAPITLAIDRANISSPGLIYTYTQDPTVTRLEPTWSIINGSTAITVSGTHLLTVQEPRVRAKYRGIETTNTCQVINDTAMLCKAPGIFLGRPQPRAQGEHPDEFGFLLDHVQTARSLNRSSFTYYPDPSFEPLGPSGVLDVKPGSHVVLKGKN | |
| 0.02 mL | |
| Growth and Development, Neuronal Cell Markers | |
| 55558 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction