Learn More
Invitrogen™ Plexin A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ 720179
Description
These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature. This antibody is predicted to react with Monkey, Cat and Pig.
Plexins are a family of large, transmembrane receptors for multiple classes of semaphorins in vertebrates. Plexins are widely expressed, and regions of their extracellular domain are homologus to both scatter factor receptors and semaphorin domains. Plexins may act as semaphorin receptors alone or in combination with neuropilins. Plexins are divided into four subfamilies designated plexin-A, -B, -C, and -D. Drosophila plexin-A is a receptor for class I semaphorins and controls motor and axon guidance. Plexin-A3 mediates cell-repelling cues. Plexins B and C are receptors for Sema 4 and Sema 7, respectively.
Specifications
| Plexin A2 | |
| Polyclonal | |
| Unconjugated | |
| PLXNA2 | |
| 2810428A13Rik; AA589422; AW457381; FLJ11751; FLJ30634; KIAA0463; LOW QUALITY PROTEIN: plexin-A2; mKIAA0463; OCT; plex 2; PlexA2; plexin 2; plexin A2; plexin-2; plexin-A2; PLXN2; Plxna2; Semaphorin receptor OCT; si:dkey-164k14.1; transmembrane protein OCT; UNQ209/PRO235 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5362 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.2 | |
| O75051 | |
| PLXNA2 | |
| Protein corresponding to Human PLXNA2 (aa 35-250). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.