missing translation for 'onlineSavingsMsg'
Learn More

Plexin A2 Antibody (583603) [FITC], Novus Biologicals™

Product Code. 30639014 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30639014 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30639014 Supplier Novus Biologicals Supplier No. FAB5486F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Plexin A2 Monoclonal antibody specifically detects Plexin A2 in Human,Mouse samples. It is validated for Western Blot,Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Plexin A2
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 583603
Conjugate FITC
Formulation PBS
Gene Alias FLJ11751, FLJ30634, KIAA0463plexin 2, OCTSemaphorin receptor OCT, plexin A2, PLXN2plexin-A2, transmembrane protein OCT
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Plexin A2, Met35-Pro1237, Accession # P70207
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5362
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.