missing translation for 'onlineSavingsMsg'
Learn More

Plexin A2 Antibody (2G5), Novus Biologicals™

Product Code. 18342509 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18342509 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18342509 Supplier Novus Biologicals Supplier No. H00005362M06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Plexin A2 Monoclonal antibody specifically detects Plexin A2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Plexin A2
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2G5
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Alias FLJ11751, FLJ30634, KIAA0463plexin 2, OCTSemaphorin receptor OCT, plexin A2, PLXN2plexin-A2, transmembrane protein OCT
Host Species Mouse
Immunogen PLXNA2 (AAH32125, 1 a.a. ~ 164 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MHSQVFSAFPYSLPLRFWVNVIKNPQFVFDIHKGSITDACLSVVAQTFMDSCSTSEHRLDKDSPSNKLLYAKDIPSYKSWVERYYADIAKLPAISDQDMNAYLAEQSRLHAVEFNMLSALNEIYSYVSKYSEELIGALEQDEQARRQRLAYKVEQLINAMSIES
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5362
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.